Learn More
Biomatik Corporation FTSJ1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08451
Encompass_Preferred
The FTSJ1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Putative tRNA (cytidine (32)/guanosine (34)-2'-O)-methyltransferase protein (1-329AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: 2'-O-ribose RNA methyltransferase TRM7 homolog Protein ftsJ homolog 1 FTSJ1 JM23. UniProt: Q9UET6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8034