Learn More
Biomatik Corporation Fxn anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08452
Encompass_Preferred
The Fxn Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant Mouse Frataxin, mitochondrial protein (78-207AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Frataxin, mitochondrial, Fxn, Frda, Fxn. UniProt: O35943. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8035