Learn More
Biomatik Corporation FYN anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08453
Encompass_Preferred
The FYN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Tyrosine-protein kinase Fyn protein (154-270AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Tyrosine-protein kinase Fyn (EC:2.7.10.2), FYN, Proto-oncogene Syn, Proto-oncogene c-Fyn, Src-like kinase, SLK, p59-Fyn. UniProt: P06241. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8036