Learn More
Biomatik Corporation G6PC anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08454
Encompass_Preferred
The G6PC Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Glucose-6-phosphatase protein (82-117AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Glucose-6-phosphatase, G-6-Pase, G6Pase, Glucose-6-phosphatase alpha, G6Pase-alpha, G6PC, G6PT. UniProt: P35575. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8037