Learn More
Biomatik Corporation GABRB2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508039
Encompass_Preferred
The GABRB2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Gamma-aminobutyric acid receptor subunit beta-2 protein (26-244AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gamma-aminobutyric acid receptor subunit beta-2, GABRB2, GABA (A) receptor subunit beta-2. UniProt: P47870. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8039 Supplier Biomatik Corporation Supplier No. CAC08456
The GABRB2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Gamma-aminobutyric acid receptor subunit beta-2 protein (26-244AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gamma-aminobutyric acid receptor subunit beta-2, GABRB2, GABA (A) receptor subunit beta-2. UniProt: P47870. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.