Learn More
Biomatik Corporation Gba anti-Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08458
Encompass_Preferred
The Gba Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: Recombinant Mouse Glucosylceramidase protein (20-515AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1:5000. Alternative Names: Glucosylceramidase, Acid beta-glucosidase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, Gba. UniProt: P17439. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8041