Learn More
Biomatik Corporation GFAP anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508046
Encompass_Preferred
The GFAP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Glial fibrillary acidic protein (292-432AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GFAP. UniProt: P14136. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8046 Supplier Biomatik Corporation Supplier No. CAC08463
The GFAP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Glial fibrillary acidic protein (292-432AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GFAP. UniProt: P14136. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.