Learn More
Biomatik Corporation GJB3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08470
Encompass_Preferred
The GJB3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Gap junction beta-3 protein (211-270AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Gap junction beta-3 protein, GJB3, CX31, Connexin-31, Cx31. UniProt: O75712. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8053