Learn More
Biomatik Corporation GLCE anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08477
Encompass_Preferred
The GLCE Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human D-glucuronyl C5-epimerase protein (29-617AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Heparan sulfate C5-epimerase, Hsepi, Heparin/heparan sulfate:glucuronic acid C5-epimerase, Heparosan-N-sulfate-glucuronate 5-epimerase, GLCE, KIAA0836. UniProt: O94923. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8060