Learn More
Biomatik Corporation GLP1R anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508062
Encompass_Preferred
The GLP1R Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Glucagon-like peptide 1 receptor protein (24-145AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GLP1R. UniProt: P43220. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8062 Supplier Biomatik Corporation Supplier No. CAC08479
The GLP1R Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Glucagon-like peptide 1 receptor protein (24-145AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: GLP1R. UniProt: P43220. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.