Learn More
Biomatik Corporation GPC4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08491
Encompass_Preferred
The GPC4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Glypican-4 protein (19-210AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Glypican-4, GPC4, UNQ474/PRO937, K-glypican, Secreted glypican-4. UniProt: O75487. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8074