Learn More
Biomatik Corporation GPT anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508079
Encompass_Preferred
The GPT Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Alanine aminotransferase 1 protein (2-496AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Glutamate pyruvate transaminase 1, Glutamic--alanine transaminase 1, Glutamic--pyruvic transaminase 1, AAT1, GPT1, GPT. UniProt: P24298. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8079 Supplier Biomatik Corporation Supplier No. CAC08496
The GPT Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Alanine aminotransferase 1 protein (2-496AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Glutamate pyruvate transaminase 1, Glutamic--alanine transaminase 1, Glutamic--pyruvic transaminase 1, AAT1, GPT1, GPT. UniProt: P24298. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.