Learn More
Biomatik Corporation HPD anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508109
Encompass_Preferred
The HPD Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human 4-hydroxyphenylpyruvate dioxygenase protein (2-393AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 4-hydroxyphenylpyruvic acid oxidase. UniProt: P32754. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8109 Supplier Biomatik Corporation Supplier No. CAC08526
The HPD Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human 4-hydroxyphenylpyruvate dioxygenase protein (2-393AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 4-hydroxyphenylpyruvic acid oxidase. UniProt: P32754. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.