Learn More
Biomatik Corporation Hpx anti-Rat, Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508110
Encompass_Preferred
The Hpx Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Human. Immunogen: Recombinant rat Hemopexin protein (24-460AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Hpx. UniProt: P20059. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8110 Supplier Biomatik Corporation Supplier No. CAC08527
The Hpx Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat, Human. Immunogen: Recombinant rat Hemopexin protein (24-460AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Hpx. UniProt: P20059. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.