Learn More
Biomatik Corporation HS2ST1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08530
Encompass_Preferred
The HS2ST1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Heparan sulfate 2-O-sulfotransferase 1 protein (29-229AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: Heparan sulfate 2-O-sulfotransferase 1 (EC:2.8.2.-), 2-O-sulfotransferase, 2OST, HS2ST1, HS2ST, KIAA0448. UniProt: Q7LGA3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8113