Learn More
Biomatik Corporation HTR1E anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08531
Encompass_Preferred
The HTR1E Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human 5-hydroxytryptamine receptor 1E protein (203-291AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 5-hydroxytryptamine receptor 1E, 5-HT-1E, 5-HT1E, S31, Serotonin receptor 1E, HTR1E. UniProt: P28566. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8114