Learn More
Biomatik Corporation IDO1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08537
Encompass_Preferred
Ido1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IDO1. The Ido1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Indoleamine 2,3-Dioxygenase 1 Protein (1-403Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IDO1. Accession Number: P14902. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8120