Learn More
Biomatik Corporation IFIT1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508122
Encompass_Preferred
Ifit1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFIT1. The Ifit1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Protein With Tetratricopeptide Repeats 1 Protein (1-181Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IFIT1. Accession Number: P09914. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8122 Supplier Biomatik Corporation Supplier No. CAC08539
Ifit1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFIT1. The Ifit1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Protein With Tetratricopeptide Repeats 1 Protein (1-181Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IFIT1. Accession Number: P09914. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.