Learn More
Biomatik Corporation IL-33 anti-Sus scrofa, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08551
Encompass_Preferred
Il-33 Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to IL-33. The Il-33 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Recombinant Pig Interleukin-33 Protein (123-276Aa). Applications: ELISA, WB. Target Synonyms: IL-33. Accession Number: M5B263. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8134