Learn More
Biomatik Corporation ACOD1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08562
Encompass_Preferred
The ACOD1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cis-aconitate decarboxylase protein (57-257AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Cis-aconitate decarboxylase, Aconitate decarboxylase, Aconitate decarboxylase, ACOD1. UniProt: A6NK06. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8145