Learn More
Biomatik Corporation KCNJ10 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08574
Encompass_Preferred
Kcnj10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KCNJ10. The Kcnj10 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Atp-Sensitive Inward Rectifier Potassium Channel 10 Protein (165-379Aa). Applications: ELISA, IHC, IF. Target Synonyms: KCNJ10. Accession Number: P78508. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8157