Learn More
Biomatik Corporation KLRK1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508164
Encompass_Preferred
Klrk1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KLRK1. The Klrk1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nkg2-D Type Ii Integral Membrane Protein (73-216Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: KLRK1. Accession Number: P26718. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8164 Supplier Biomatik Corporation Supplier No. CAC08581
Klrk1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KLRK1. The Klrk1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nkg2-D Type Ii Integral Membrane Protein (73-216Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: KLRK1. Accession Number: P26718. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.