Learn More
Biomatik Corporation LIPA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508176
Encompass_Preferred
Lipa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LIPA. The Lipa Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lysosomal Acid Lipase; Cholesteryl Ester Hydrolase Protein (22-399Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LIPA. Accession Number: P38571. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8176 Supplier Biomatik Corporation Supplier No. CAC08593
Lipa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LIPA. The Lipa Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lysosomal Acid Lipase; Cholesteryl Ester Hydrolase Protein (22-399Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LIPA. Accession Number: P38571. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.