Learn More
Biomatik Corporation MDM4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508203
Encompass_Preferred
Mdm4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MDM4. The Mdm4 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Mdm4 Protein (208-306Aa). Applications: ELISA, WB. Target Synonyms: MDM4. Accession Number: O15151. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8203 Supplier Biomatik Corporation Supplier No. CAC08620
Mdm4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MDM4. The Mdm4 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Mdm4 Protein (208-306Aa). Applications: ELISA, WB. Target Synonyms: MDM4. Accession Number: O15151. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.