Learn More
Biomatik Corporation NQO2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08680
Encompass_Preferred
Nqo2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NQO2. The Nqo2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ribosyldihydronicotinamide Dehydrogenase [Quinone] Protein (1-231Aa). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: NQO2. Accession Number: P16083. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8263