Learn More
Biomatik Corporation NRP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508267
Encompass_Preferred
Nrp2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NRP2. The Nrp2 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neuropilin-2 Protein (60-209Aa). Applications: ELISA, IHC, IF. Target Synonyms: NRP2. Accession Number: O60462. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8267 Supplier Biomatik Corporation Supplier No. CAC08684
Nrp2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NRP2. The Nrp2 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Neuropilin-2 Protein (60-209Aa). Applications: ELISA, IHC, IF. Target Synonyms: NRP2. Accession Number: O60462. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.