Learn More
Biomatik Corporation OVOL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08700
Encompass_Preferred
Ovol1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to OVOL1. The Ovol1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Putative Transcription Factor Ovo-Like 1 Protein (9-267Aa). Applications: ELISA, WB, IHC. Target Synonyms: OVOL1. Accession Number: O14753. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8283