Learn More
Biomatik Corporation PAH anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508286
Encompass_Preferred
Pah Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PAH. The Pah Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phenylalanine-4-Hydroxylase Protein (2-452Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PAH. Accession Number: P00439. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8286 Supplier Biomatik Corporation Supplier No. CAC08703
Pah Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PAH. The Pah Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phenylalanine-4-Hydroxylase Protein (2-452Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PAH. Accession Number: P00439. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.