Learn More
Biomatik Corporation BUD31 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08706
Encompass_Preferred
The BUD31 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein BUD31 homolog protein (1-144AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Protein BUD31 homolog, Protein EDG-2, Protein G10 homolog, BUD31, EDG2. UniProt: P41223. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8289