Learn More
Biomatik Corporation def anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508295
Encompass_Preferred
The def Polyclonal Antibody is raised in Rabbit. Species Reactivity: Escherichia coli. Immunogen: Recombinant Escherichia coli Peptide deformylase protein (2-169AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: PDF, Polypeptide deformylase, b3287, JW3248, fms, def. UniProt: P0A6K3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8295 Supplier Biomatik Corporation Supplier No. CAC08712
The def Polyclonal Antibody is raised in Rabbit. Species Reactivity: Escherichia coli. Immunogen: Recombinant Escherichia coli Peptide deformylase protein (2-169AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: PDF, Polypeptide deformylase, b3287, JW3248, fms, def. UniProt: P0A6K3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.