Learn More
Biomatik Corporation PFKFB2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08717
Encompass_Preferred
Pfkfb2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PFKFB2. The Pfkfb2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 6-Phosphofructo-2-Kinase; Fructose-2,6-Bisphosphatase 2 Protein (360-505Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PFKFB2. Accession Number: O60825. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8300