Learn More
Biomatik Corporation PGP anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08719
Encompass_Preferred
Pgp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PGP. The Pgp Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Glycerol-3-Phosphate Phosphatase Protein (140-300Aa). Applications: ELISA, IHC, IF. Target Synonyms: PGP. Accession Number: A6NDG6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8302