Learn More
Biomatik Corporation Plau anti-Rat, Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508311
Encompass_Preferred
Plaur Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Plaur. The Plaur Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Recombinant Rat Urokinase Plasminogen Activator Surface Receptor Ptotein (25-299Aa). Applications: ELISA, WB. Target Synonyms: Plaur. Accession Number: P49616. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8311 Supplier Biomatik Corporation Supplier No. CAC08728
Plaur Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Plaur. The Plaur Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Recombinant Rat Urokinase Plasminogen Activator Surface Receptor Ptotein (25-299Aa). Applications: ELISA, WB. Target Synonyms: Plaur. Accession Number: P49616. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.