Learn More
Biomatik Corporation PLPP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08744
Encompass_Preferred
Plpp2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLPP2. The Plpp2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phospholipid Phosphatase 2 Protein (109-162Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PLPP2. Accession Number: O43688. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8327