Learn More
Biomatik Corporation PRAME anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08754
Encompass_Preferred
Prame Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRAME. The Prame Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Melanoma Antigen Preferentially Expressed In Tumors Protein (1-509Aa). Applications: ELISA, IHC. Target Synonyms: PRAME. Accession Number: P78395. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8337