Learn More
Biomatik Corporation Prf1 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508338
Encompass_Preferred
Prf1 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Prf1. The Prf1 Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Perforin-1 Protein (21-554Aa). Applications: ELISA. Target Synonyms: Prf1. Accession Number: P10820. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8338 Supplier Biomatik Corporation Supplier No. CAC08755
Prf1 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Prf1. The Prf1 Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Perforin-1 Protein (21-554Aa). Applications: ELISA. Target Synonyms: Prf1. Accession Number: P10820. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.