Learn More
Biomatik Corporation THAP12 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08756
Encompass_Preferred
Thap12 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to THAP12. The Thap12 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 52 Kda Repressor Of The Inhibitor Of The Protein Kinase Protein (612-761Aa). Applications: ELISA, WB, IHC. Target Synonyms: THAP12. Accession Number: O43422. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8339