Learn More
Biomatik Corporation PSMA2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08763
Encompass_Preferred
Psma2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PSMA2. The Psma2 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Proteasome Subunit Alpha Type-2 Protein (120-227Aa). Applications: ELISA, IHC, IF. Target Synonyms: PSMA2. Accession Number: P25787. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8346