Learn More
Biomatik Corporation RAB27A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08772
Encompass_Preferred
Rab27A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAB27A. The Rab27A Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related Protein Rab-27A Protein (2-221Aa). Applications: ELISA, IHC, IF. Target Synonyms: RAB27A. Accession Number: P51159. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8355