Learn More
Biomatik Corporation RAB4B anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08774
Encompass_Preferred
Rab4B Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RAB4B. The Rab4B Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ras-Related Protein Rab-4B Protein (2-213Aa). Applications: ELISA, WB, IHC. Target Synonyms: RAB4B. Accession Number: P61018. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8357