Learn More
Biomatik Corporation RBM10 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508367
Encompass_Preferred
Rbm10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RBM10. The Rbm10 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Rna-Binding Protein 10 Protein (501-700Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RBM10. Accession Number: P98175. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8367 Supplier Biomatik Corporation Supplier No. CAC08784
Rbm10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RBM10. The Rbm10 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Rna-Binding Protein 10 Protein (501-700Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RBM10. Accession Number: P98175. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.