Learn More
Biomatik Corporation RNASE2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08801
Encompass_Preferred
Rnase2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RNASE2. The Rnase2 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Non-Secretory Ribonuclease Protein (28-161Aa). Applications: ELISA, IHC. Target Synonyms: RNASE2. Accession Number: P10153. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8384