Learn More
Biomatik Corporation RPS8 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08814
Encompass_Preferred
Rps8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RPS8. The Rps8 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 40S Ribosomal Protein S8 Protein (2-208Aa). Applications: ELISA, WB. Target Synonyms: RPS8. Accession Number: P62241. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8397