Learn More
Biomatik Corporation RTEL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08820
Encompass_Preferred
Rtel1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RTEL1. The Rtel1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Regulator Of Telomere Elongation Helicase 1 Protein (975-1219Aa). Applications: ELISA, IHC, IF. Target Synonyms: RTEL1. Accession Number: Q9NZ71. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8403