Learn More
Biomatik Corporation SCGB1A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08828
Encompass_Preferred
Scgb1A1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCGB1A1. The Scgb1A1 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Uteroglobin Protein (22-91Aa). Applications: ELISA, IF. Target Synonyms: SCGB1A1. Accession Number: P11684. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8411