Learn More
Biomatik Corporation PMEL anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508428
Encompass_Preferred
Pmel Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PMEL. The Pmel Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Melanocyte Protein Pmel Protein (25-467Aa). Applications: ELISA, WB, IHC. Target Synonyms: PMEL. Accession Number: P40967. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8428 Supplier Biomatik Corporation Supplier No. CAC08845
Pmel Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PMEL. The Pmel Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Melanocyte Protein Pmel Protein (25-467Aa). Applications: ELISA, WB, IHC. Target Synonyms: PMEL. Accession Number: P40967. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.