Learn More
Biomatik Corporation SIRPA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508429
Encompass_Preferred
Sirpa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SIRPA. The Sirpa Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tyrosine-Protein Phosphatase Non-Receptor Type Substrate 1 Protein (31-373Aa). Applications: ELISA, IHC, IF. Target Synonyms: SIRPA. Accession Number: P78324. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8429 Supplier Biomatik Corporation Supplier No. CAC08846
Sirpa Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SIRPA. The Sirpa Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tyrosine-Protein Phosphatase Non-Receptor Type Substrate 1 Protein (31-373Aa). Applications: ELISA, IHC, IF. Target Synonyms: SIRPA. Accession Number: P78324. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.