Learn More
Biomatik Corporation SLC1A6 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08848
Encompass_Preferred
Slc1A6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC1A6. The Slc1A6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Excitatory Amino Acid Transporter 4 Protein (149-271Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC1A6. Accession Number: P48664. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8431