Learn More
Biomatik Corporation SNX20 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08862
Encompass_Preferred
Snx20 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SNX20. The Snx20 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sorting Nexin-20 Protein (1-102Aa). Applications: ELISA, WB, IHC. Target Synonyms: SNX20. Accession Number: Q7Z614. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8445