Learn More
Biomatik Corporation SOCS3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08863
Encompass_Preferred
Socs3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SOCS3. The Socs3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Suppressor Of Cytokine Signaling 3 Protein (1-224Aa). Applications: ELISA, IHC, IF. Target Synonyms: SOCS3. Accession Number: O14543. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8446